Gene Transcripts

Gene Symbol
MIR17HG
Entrez Gene ID
407975
Gene Name
miR-17-92a-1 cluster host gene
Gene Aliases
C13orf25, FGLDS2, LINC00048, MIHG1, MIRH1, MIRHG1, NCRNA00048, miR-17-92
Chromosome Location
Chr. 13: 91347820 - 91354575 on Build GRCh38
UniGene ID
Hs.24115
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027349.1 4 843
NR_027350.1 2 4934
GenBank mRNA AK024240.1 - 1 2657

Gene Symbol
MIR18A
Entrez Gene ID
406953
Gene Name
microRNA 18a
Gene Aliases
MIR18, MIRN18, MIRN18A, hsa-mir-18, hsa-mir-18a, miR-18, miRNA18A, mir-18a
Chromosome Location
Chr. 13: 91350751 - 91350821 on Build GRCh38
UniGene ID
Hs.24115
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB176708.1 - LIT3364
oncomir-1
2 4934

View Full Product Details