Gene Transcripts
- Gene Symbol
- NOC2LP2
- Entrez Gene ID
- 401010
- Gene Name
- NOC2 like nucleolar associated transcriptional repressor pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 131442161 - 131444894 on Build GRCh38
- UniGene ID
- Hs.452369
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002826.2 |
|
1 |
2232 |
|
GenBank mRNA |
AK302229.1 |
- |
1 |
2232 |
|
BC060851.1 |
- |
1 |
2225 |
|
View Full Product Details