Gene Transcripts
- Gene Symbol
- PSCA
- Entrez Gene ID
- 8000
- Gene Name
- prostate stem cell antigen
- Gene Aliases
- PRO232
- Chromosome Location
- Chr. 8: 142670308 - 142682727 on Build GRCh38
- UniGene ID
- Hs.652235
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033343.1 |
|
1 |
99 |
|
GenBank mRNA |
BI758811.1 |
- |
1 |
97 |
|
BU157227.1 |
- |
1 |
86 |
|
View Full Product Details