Gene Transcripts
- Gene Symbol
- PTTG3P
- Entrez Gene ID
- 26255
- Gene Name
- pituitary tumor-transforming 3, pseudogene
- Gene Aliases
- PTTG3, rcPTTG1
- Chromosome Location
- Chr. 8: 66767397 - 66768005 on Build GRCh38
- UniGene ID
- Hs.647156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002734.2 |
|
1 |
155 |
|
GenBank mRNA |
AF095289.1 |
- |
1 |
126 |
|
View Full Product Details