Gene Transcripts

Gene Symbol
NEAT1
Entrez Gene ID
283131
Gene Name
nuclear paraspeckle assembly transcript 1 (non-protein coding)
Gene Aliases
LINC00084, NCRNA00084, TncRNA, VINC
Chromosome Location
Chr. 11: 65422798 - 65445540 on Build GRCh38
UniGene ID
Hs.523789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028272.1 1 753
NR_131012.1 1 753
GenBank mRNA EF177379.1 - 1 753
GQ859162.1 - 1 753


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK026198.1 - 1 747 Chr. 11 - 65422804 - 65423809 [+]

View Full Product Details