Gene Transcripts
- Gene Symbol
- RPL19P12
- Entrez Gene ID
- 100129424
- Gene Name
- ribosomal protein L19 pseudogene 12
- Gene Aliases
- RPL19_4_829
- Chromosome Location
- Chr. 7: 103141270 - 103142403 on Build GRCh38
- UniGene ID
- Hs.720729
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026660.1 |
|
1 |
296 |
|
GenBank mRNA |
AW373666.1 |
- |
1 |
298 |
|
CD709306.1 |
- |
1 |
299 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW373669.1 |
- |
1 |
296 |
|
Chr. 7 - 103141771 - 103142399 [-] |
GenBankmRNA |
HY113687.1 |
- |
1 |
324 |
|
Chr. 7 - 103141963 - 103142431 [-] |
View Full Product Details