Gene Transcripts

Gene Symbol
D2Ertd173e
Entrez Gene ID
51864
Gene Name
DNA segment, Chr 2, ERATO Doi 173, expressed
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK083486.1 - LIT1672
Nespas
1 1279

View Full Product Details