Gene Transcripts

Gene Symbol
OR5AK4P
Entrez Gene ID
219525
Gene Name
olfactory receptor family 5 subfamily AK member 4 pseudogene
Gene Aliases
PJCG3
Chromosome Location
Chr. 11: 57037534 - 57038460 on Build GRCh38
UniGene ID
Hs.554519
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_036445.1
1 436
GenBank mRNA AF309700.1 - LIT2083
PJCG3 pseudogene
1 86

View Full Product Details