Gene Transcripts

Gene Symbol
MGST1
Entrez Gene ID
4257
Gene Name
microsomal glutathione S-transferase 1
Gene Aliases
GST12, MGST, MGST-I
Chromosome Location
Chr. 12: 16347142 - 16377189 on Build GRCh38
UniGene ID
Hs.389700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK123482.1 - 1 1139

View Full Product Details