Gene Transcripts

Gene Symbol
TRIM14
Entrez Gene ID
9830
Gene Name
tripartite motif containing 14
Gene Aliases
Chromosome Location
Chr. 9: 98072014 - 98119418 on Build GRCh38
UniGene ID
Hs.575631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK123739.1 - 1 145

View Full Product Details