Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286238.1 | NP_001273167.1 | 13 | 4445 | |
NM_001286239.1 | NP_001273168.1 | 12 | 4451 | ||
NM_019073.3 | NP_061946.1 | 13 | 4493 | ||
XM_006710700.2 | XP_006710763.1 | 11 | 4292 | ||
XM_011541606.1 | XP_011539908.1 | 11 | 4202 | ||
XM_011541607.2 | XP_011539909.1 | 10 | 4056 | ||
XM_011541608.1 | XP_011539910.1 | 12 | 4430 | ||
XM_011541609.2 | XP_011539911.1 | 11 | 4138 | ||
XM_017001510.1 | XP_016856999.1 | 10 | 4242 | ||
XM_017001511.1 | XP_016857000.1 | 12 | 4395 | ||
XM_017001512.1 | XP_016857001.1 | 14 | 4336 | ||
XM_017001513.1 | XP_016857002.1 | 11 | 4100 | ||
XM_017001514.1 | XP_016857003.1 | 14 | 4378 | ||
XM_017001515.1 | XP_016857004.1 | 12 | 4223 | ||
GenBank mRNA | BC040655.1 | - | 1 | 904 |