Gene Transcripts
- Gene Symbol
- HHLA2
- Entrez Gene ID
- 11148
- Gene Name
- HERV-H LTR-associating 2
- Gene Aliases
- B7-H5, B7-H7, B7H7, B7y
- Chromosome Location
- Chr. 3: 108296490 - 108378285 on Build GRCh38
- UniGene ID
- Hs.225968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097806.1 |
- |
1 |
1120 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB213093.1 |
- |
1 |
135 |
|
Chr. 3 - 108388581 - 108389134 [+] |
View Full Product Details