Gene Transcripts

Gene Symbol
TSNAX-DISC1
Entrez Gene ID
100303453
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 1: 231528653 - 232041272 on Build GRCh38
UniGene ID
Hs.13318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF230316.1 - 1 158

View Full Product Details