Gene Transcripts

Gene Symbol
NDE1
Entrez Gene ID
54820
Gene Name
nudE neurodevelopment protein 1
Gene Aliases
HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1
Chromosome Location
Chr. 16: 15643267 - 15726353 on Build GRCh38
UniGene ID
Hs.655378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017023349.1 XP_016878838.1 9 3110
XM_017023351.1 XP_016878840.1 10 3189
XM_017023353.1 XP_016878842.1 9 3242
XM_017023354.1 XP_016878843.1 8 2958
XM_017023356.1 XP_016878845.1 9 3146
GenBank mRNA AK123247.1 - 2 2203


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU935437.1 - 262 Chr. - - [-]
GenBankmRNA CA941555.1 - 1 510 Chr. 16 - 15701280 - 15701837 [+]
GenBankmRNA BP431596.1 - 1 310 Chr. 16 - 15701480 - 15701850 [+]
GenBankmRNA BG112775.1 - 1 454 Chr. 16 - 15701332 - 15701913 [+]

View Full Product Details