Gene Transcripts
- Gene Symbol
- BLACAT1
- Entrez Gene ID
- 101669762
- Gene Name
- bladder cancer associated transcript 1 (non-protein coding)
- Gene Aliases
- LINC00912, linc-UBC1, onco-lncRNA-30
- Chromosome Location
- Chr. 1: 205434886 - 205456086 on Build GRCh38
- UniGene ID
- Hs.666219
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_103783.1 |
|
2 |
2382 |
|
GenBank mRNA |
AK022594.1 |
- |
1 |
2135 |
|
BX640973.1 |
- |
1 |
2763 |
|
View Full Product Details