Gene Transcripts

Gene Symbol
RRP1
Entrez Gene ID
8568
Gene Name
ribosomal RNA processing 1
Gene Aliases
D21S2056E, NNP-1, NOP52, RRP1A
Chromosome Location
Chr. 21: 43789537 - 43804102 on Build GRCh38
UniGene ID
Hs.110757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017028485.1 XP_016883974.1 13 1339
GenBank mRNA AK026354.1 - 1 237


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW901603.1 - 1 422 Chr. 21 - 43803045 - 43803536 [+]
GenBankmRNA BP263582.1 - 1 237 Chr. 21 - 43803216 - 43803797 [+]
GenBankmRNA BF363881.1 - 1 375 Chr. 21 - 43803083 - 43803555 [+]

View Full Product Details