Gene Transcripts

Gene Symbol
RASGRF1
Entrez Gene ID
5923
Gene Name
Ras protein specific guanine nucleotide releasing factor 1
Gene Aliases
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
Chromosome Location
Chr. 15: 78959947 - 79090873 on Build GRCh38
UniGene ID
Hs.459035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL359931.1 - 1 1468


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA119282.1 - 1 189 Chr. 15 - 78979837 - 78980387 [-]

View Full Product Details