Gene Transcripts
- Gene Symbol
- C22orf23
- Entrez Gene ID
- 84645
- Gene Name
- chromosome 22 open reading frame 23
- Gene Aliases
- EVG1, dJ1039K5.6
- Chromosome Location
- Chr. 22: 37943050 - 37953669 on Build GRCh38
- UniGene ID
- Hs.517612
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DA968042.1 |
- |
1 |
375 |
|
- Gene Symbol
- MICALL1
- Entrez Gene ID
- 85377
- Gene Name
- MICAL like 1
- Gene Aliases
- MICAL-L1, MIRAB13
- Chromosome Location
- Chr. 22: 37906148 - 37942458 on Build GRCh38
- UniGene ID
- Hs.517610
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB051455.1 |
- |
14 |
4969 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK097339.1 |
- |
1 |
375 |
|
Chr. 22 - 37943050 - 37944898 [+] |
View Full Product Details