Gene Transcripts

Gene Symbol
PANK3
Entrez Gene ID
79646
Gene Name
pantothenate kinase 3
Gene Aliases
Chromosome Location
Chr. 5: 168555623 - 168579609 on Build GRCh38
UniGene ID
Hs.388400
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M55536.1 - 1 1209


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA803138.1 - 1 299 Chr. 5 - 168553149 - 168553706 [-]
GenBankmRNA DA803157.1 - 1 299 Chr. 5 - 168553142 - 168553706 [-]
GenBankmRNA DA803120.1 - 1 299 Chr. 5 - 168553152 - 168553706 [-]
GenBankmRNA DA803108.1 - 1 299 Chr. 5 - 168553151 - 168553706 [-]
GenBankmRNA DA803121.1 - 1 299 Chr. 5 - 168553154 - 168553706 [-]
GenBankmRNA DA803109.1 - 1 299 Chr. 5 - 168553150 - 168553706 [-]
GenBankmRNA DA803132.1 - 1 299 Chr. 5 - 168553166 - 168553706 [-]
GenBankmRNA DA803114.1 - 1 299 Chr. 5 - 168553153 - 168553706 [-]
GenBankmRNA DA803152.1 - 1 299 Chr. 5 - 168553140 - 168553706 [-]
GenBankmRNA DB150431.1 - 1 502 Chr. 5 - 168553342 - 168553908 [-]
GenBankmRNA DA803146.1 - 1 299 Chr. 5 - 168553192 - 168553706 [-]
GenBankmRNA DA803102.1 - 1 299 Chr. 5 - 168553149 - 168553706 [-]
GenBankmRNA BF895978.1 - 1 234 Chr. 5 - 168553240 - 168553631 [-]
GenBankmRNA DA803126.1 - 1 299 Chr. 5 - 168553163 - 168553706 [-]
GenBankmRNA DA803127.1 - 1 299 Chr. 5 - 168553151 - 168553706 [-]

View Full Product Details