Gene Transcripts

Gene Symbol
SLC5A9
Entrez Gene ID
200010
Gene Name
solute carrier family 5 member 9
Gene Aliases
SGLT4
Chromosome Location
Chr. 1: 48221113 - 48248644 on Build GRCh38
UniGene ID
Hs.37890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC032417.1 - 1 1403

View Full Product Details