Gene Transcripts

Gene Symbol
ZCCHC7
Entrez Gene ID
84186
Gene Name
zinc finger CCHC-type containing 7
Gene Aliases
AIR1, HSPC086
Chromosome Location
Chr. 9: 37120169 - 37358149 on Build GRCh38
UniGene ID
Hs.654700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC022434.1 - 4 905


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK126219.1 - 1 2991 Chr. 9 - 37183751 - 37186999 [+]

View Full Product Details