Gene Transcripts
- Gene Symbol
- LBHD1
- Entrez Gene ID
- 79081
- Gene Name
- LBH domain containing 1
- Gene Aliases
- C11orf48
- Chromosome Location
- Chr. 11: 62662817 - 62671769 on Build GRCh38
- UniGene ID
- Hs.9061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124145.1 |
- |
1 |
1082 |
|
- Gene Symbol
- C11orf98
- Entrez Gene ID
- 102288414
- Gene Name
- chromosome 11 open reading frame 98
- Gene Aliases
- C11orf48
- Chromosome Location
- Chr. 11: 62662817 - 62665216 on Build GRCh38
- UniGene ID
- Hs.9061
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104414.1 |
|
1 |
110 |
|
GenBank mRNA |
BG681685.1 |
- |
1 |
54 |
|
View Full Product Details