Gene Transcripts
- Gene Symbol
- SLC5A3
- Entrez Gene ID
- 6526
- Gene Name
- solute carrier family 5 member 3
- Gene Aliases
- BCW2, SMIT, SMIT1, SMIT2
- Chromosome Location
- Chr. 21: 34073523 - 34106262 on Build GRCh38
- UniGene ID
- Hs.302742
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055913.1 |
- |
1 |
2086 |
|
View Full Product Details