Gene Transcripts
- Gene Symbol
- TNRC18
- Entrez Gene ID
- 84629
- Gene Name
- trinucleotide repeat containing 18
- Gene Aliases
- CAGL79, TNRC18A
- Chromosome Location
- Chr. 7: 5306790 - 5426043 on Build GRCh38
- UniGene ID
- Hs.520638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY129022.1 |
- |
1 |
1153 |
|
View Full Product Details