Gene Transcripts

Gene Symbol
VN2R9P
Entrez Gene ID
648315
Gene Name
vomeronasal 2 receptor 9 pseudogene
Gene Aliases
Chromosome Location
Chr. 11: 58040768 - 58066959 on Build GRCh38
UniGene ID
Hs.652053
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC067882.1 - 4 1254

View Full Product Details