Gene Transcripts
- Gene Symbol
- NAF1
- Entrez Gene ID
- 92345
- Gene Name
- nuclear assembly factor 1 ribonucleoprotein
- Gene Aliases
- Chromosome Location
- Chr. 4: 163109133 - 163170388 on Build GRCh38
- UniGene ID
- Hs.129095
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126037.1 |
- |
1 |
524 |
|
View Full Product Details