Gene Transcripts

Gene Symbol
BCL2L13
Entrez Gene ID
23786
Gene Name
BCL2 like 13
Gene Aliases
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location
Chr. 22: 17628855 - 17730855 on Build GRCh38
UniGene ID
Hs.631672
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF086412.1 - 1 473


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX099464.1 - 1 473 Chr. 22 - 17706456 - 17706990 [+]

View Full Product Details