Gene Transcripts
- Gene Symbol
- SIM1
- Entrez Gene ID
- 6492
- Gene Name
- single-minded family bHLH transcription factor 1
- Gene Aliases
- bHLHe14
- Chromosome Location
- Chr. 6: 100385009 - 100464929 on Build GRCh38
- UniGene ID
- Hs.520293
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB522103.1 |
- |
1 |
360 |
|
Chr. 6 - 100385016 - 100385496 [-] |
GenBankmRNA |
AL049965.1 |
- |
1 |
215 |
|
Chr. 6 - 100385015 - 100385349 [-] |
GenBankmRNA |
AL036129.2 |
- |
1 |
253 |
|
Chr. 6 - 100385015 - 100385349 [-] |
View Full Product Details