Gene Transcripts
- Gene Symbol
- STRN
- Entrez Gene ID
- 6801
- Gene Name
- striatin
- Gene Aliases
- PPP2R6A, SG2NA
- Chromosome Location
- Chr. 2: 36837698 - 36966607 on Build GRCh38
- UniGene ID
- Hs.127486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC036416.1 |
- |
1 |
668 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG707422.1 |
- |
1 |
669 |
|
Chr. 2 - 36851696 - 36852512 [-] |
View Full Product Details