Gene Transcripts
- Gene Symbol
- OR7E24
- Entrez Gene ID
- 26648
- Gene Name
- olfactory receptor family 7 subfamily E member 24
- Gene Aliases
- HSHT2, OR19-8, OR7E24P, OR7E24Q
- Chromosome Location
- Chr. 19: 9251044 - 9252063 on Build GRCh38
- UniGene ID
- Hs.129832
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC146920.1 |
- |
1 |
1172 |
|
Y10529.1 |
- |
1 |
833 |
|
View Full Product Details