Gene Transcripts

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
Chromosome Location
Chr. 3: 138187248 - 138298389 on Build GRCh38
UniGene ID
Hs.744868
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK128702.1 - 1 2567


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN293842.1 - 1 325 Chr. 3 - 138261036 - 138261651 [+]

View Full Product Details