Gene Transcripts

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
Chromosome Location
Chr. 2: 210002565 - 210029156 on Build GRCh38
UniGene ID
Hs.282260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006712677.3 XP_006712740.1 7 4715
GenBank mRNA AK055724.1 - 1 1369

View Full Product Details