Gene Transcripts

Gene Symbol
OXR1
Entrez Gene ID
55074
Gene Name
oxidation resistance 1
Gene Aliases
Nbla00307, TLDC3
Chromosome Location
Chr. 8: 106270178 - 106752694 on Build GRCh38
UniGene ID
Hs.127286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK098168.1 - 1 152


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB203805.1 - 1 152 Chr. 8 - 106749730 - 106750292 [+]

View Full Product Details