Gene Transcripts
- Gene Symbol
- Dep1
- Entrez Gene ID
- 170624
- Gene Name
- diabetic embryopathy 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.406761
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF032130.1 |
- |
1 |
1263 |
|
View Full Product Details