Gene Transcripts
- Gene Symbol
- Rab3gap1
- Entrez Gene ID
- 226407
- Gene Name
- RAB3 GTPase activating protein subunit 1
- Gene Aliases
- 1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130
- Chromosome Location
- Chr. 1: 127868737 - 127943876 on Build GRCm38
- UniGene ID
- Mm.489713
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005623.1 |
- |
1 |
390 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY705821.1 |
- |
1 |
390 |
|
Chr. 1 - 127938379 - 127938882 [-] |
View Full Product Details