Gene Transcripts
- Gene Symbol
- Mif
- Entrez Gene ID
- 17319
- Gene Name
- macrophage migration inhibitory factor
- Gene Aliases
- GIF, Glif
- Chromosome Location
- Chr. 10: 75859353 - 75860250 on Build GRCm38
- UniGene ID
- Mm.2326
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015344.1 |
- |
|
1231 |
|
View Full Product Details