Gene Transcripts
- Gene Symbol
- Tmem14a
- Entrez Gene ID
- 75712
- Gene Name
- transmembrane protein 14A
- Gene Aliases
- 5730496E24Rik, C85783, PTD011
- Chromosome Location
- Chr. 1: 21166436 - 21230167 on Build GRCm38
- UniGene ID
- Mm.304763
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138554.1 |
- |
2 |
317 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB633564.1 |
- |
2 |
317 |
|
Chr. 1 - 21166498 - 21187631 [+] |
View Full Product Details