Gene Transcripts
- Gene Symbol
- Tbx20
- Entrez Gene ID
- 57246
- Gene Name
- T-box 20
- Gene Aliases
- 9430010M06Rik, AL022859, Tbx12
- Chromosome Location
- Chr. 9: 24720812 - 24774303 on Build GRCm38
- UniGene ID
- Mm.31452
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK145503.1 |
- |
1 |
1676 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF902156.1 |
- |
1 |
572 |
|
Chr. 9 - 24721952 - 24722608 [-] |
View Full Product Details