Gene Transcripts
- Gene Symbol
- Cstf3
- Entrez Gene ID
- 228410
- Gene Name
- cleavage stimulation factor, 3' pre-RNA, subunit 3
- Gene Aliases
- 4732468G05Rik, C79532, CstF-77
- Chromosome Location
- Chr. 2: 104590484 - 104665425 on Build GRCm38
- UniGene ID
- Mm.259876
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK158169.1 |
- |
1 |
2406 |
|
View Full Product Details