Gene Transcripts

Gene Symbol
Gpc4
Entrez Gene ID
14735
Gene Name
glypican 4
Gene Aliases
9530073D23Rik, AF311610, AI385680, AI661372, K-glypican
Chromosome Location
Chr. X: 52050885 - 52164923 on Build GRCm38
UniGene ID
Mm.1528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK086914.1 - 1 379
AK142313.1 - 1 1017
AK153781.1 - 1 881


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BB605463.2 - 1 377 Chr. X - 52051386 - 52052034 [-]
GenBankmRNA BB530689.2 - 1 410 Chr. X - 52051373 - 52052028 [-]
GenBankmRNA CN683618.1 - 1 438 Chr. X - 52051566 - 52052095 [-]
GenBankmRNA CB320196.1 - 407 Chr. - - [-]

View Full Product Details