Gene Transcripts
- Gene Symbol
- Dusp3
- Entrez Gene ID
- 72349
- Gene Name
- dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related)
- Gene Aliases
- 2210015O03Rik, 5031436O03Rik, T-DSP11, VHR
- Chromosome Location
- Chr. 11: 101971141 - 101987005 on Build GRCm38
- UniGene ID
- Mm.196295
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077266.1 |
- |
1 |
916 |
|
View Full Product Details