Gene Transcripts

Gene Symbol
Klhdc8b
Entrez Gene ID
78267
Gene Name
kelch domain containing 8B
Gene Aliases
4931406O17Rik
Chromosome Location
Chr. 9: 108447636 - 108461581 on Build GRCm38
UniGene ID
Mm.283194
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK157991.1 - 1 1311
AK161517.1 - 1 460


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ047877.1 - 1 460 Chr. 9 - 108459240 - 108460078 [-]

View Full Product Details