Gene Transcripts
- Gene Symbol
- PDE12
- Entrez Gene ID
- 201626
- Gene Name
- phosphodiesterase 12
- Gene Aliases
- 2'-PDE, 2-PDE
- Chromosome Location
- Chr. 3: 57556247 - 57656480 on Build GRCh38
- UniGene ID
- Hs.572993
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG208290.1 |
- |
1 |
273 |
|
Chr. 3 - 57654632 - 57654947 [+] |
GenBankmRNA |
DB506931.1 |
- |
3 |
433 |
|
Chr. 3 - 57557628 - 57654944 [+] |
GENCODE |
ENST00000470427.1 |
- |
2 |
347 |
|
Chr. 3 - 57628810 - 57654918 [+] |
GenBankmRNA |
BG218317.1 |
- |
1 |
276 |
|
Chr. 3 - 57654626 - 57654979 [+] |
View Full Product Details