Gene Transcripts

Gene Symbol
PLEKHG4B
Entrez Gene ID
153478
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Gene Aliases
Chromosome Location
Chr. 5: 54065 - 189972 on Build GRCh38
UniGene ID
Hs.535800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006714446.3 XP_006714509.1 1, 2 169
XM_011513968.2 XP_011512270.1 1, 2 169
XM_011513969.2 XP_011512271.1 1, 2 169
XM_011513972.2 XP_011512274.1 1, 2 170
XM_011513973.1 XP_011512275.1 1, 2 167
XM_017009112.1 XP_016864601.1 1, 2 170

View Full Product Details