Gene Transcripts
- Gene Symbol
- MIAT
- Entrez Gene ID
- 440823
- Gene Name
- myocardial infarction associated transcript (non-protein coding)
- Gene Aliases
- C22orf35, GOMAFU, LINC00066, NCRNA00066, RNCR2, lncRNA-MIAT
- Chromosome Location
- Chr. 22: 26657482 - 26676478 on Build GRCh38
- UniGene ID
- Hs.517502
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000620145.4 |
- |
|
7289 |
|
Chr. - - [-] |
GENCODE |
ENST00000613780.4 |
- |
|
7363 |
|
Chr. - - [-] |
GENCODE |
ENST00000616469.4 |
- |
|
7289 |
|
Chr. - - [-] |
GENCODE |
ENST00000616213.4 |
- |
|
7163 |
|
Chr. - - [-] |
View Full Product Details