Gene Transcripts
- Gene Symbol
- FAM66C
- Entrez Gene ID
- 440078
- Gene Name
- family with sequence similarity 66 member C
- Gene Aliases
- Chromosome Location
- Chr. 12: 8180209 - 8201000 on Build GRCh38
- UniGene ID
- Hs.512257
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026788.1 |
|
2 |
1004 |
|
GenBank mRNA |
AK096301.1 |
- |
2 |
1004 |
|
BC034777.1 |
- |
2 |
944 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000454799.6 |
- |
2 |
1004 |
|
Chr. 12 - 8180209 - 8216151 [+] |
GENCODE |
ENST00000541558.5 |
- |
2 |
944 |
|
Chr. 12 - 8180269 - 8199881 [+] |
View Full Product Details