Gene Transcripts
- Gene Symbol
- DCAF8
- Entrez Gene ID
- 50717
- Gene Name
- DDB1 and CUL4 associated factor 8
- Gene Aliases
- GAN2, H326, WDR42A
- Chromosome Location
- Chr. 1: 160215715 - 160262560 on Build GRCh38
- UniGene ID
- Hs.632447
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026213.1 |
- |
1 |
1013 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000442130.1 |
- |
2 |
528 |
|
Chr. 1 - 160261744 - 160262778 [+] |
View Full Product Details