Gene Transcripts
- Gene Symbol
- PITRM1-AS1
- Entrez Gene ID
- 100507034
- Gene Name
- PITRM1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 3141601 - 3148629 on Build GRCh38
- UniGene ID
- Hs.665365
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038284.1 |
|
4 |
628 |
|
GenBank mRNA |
AK097545.1 |
- |
3 |
378 |
|
BC050332.1 |
- |
4 |
553 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000430356.3 |
- |
3 |
557 |
|
Chr. 10 - 3143049 - 3148622 [+] |
View Full Product Details