Gene Transcripts

Gene Symbol
DNAJB8-AS1
Entrez Gene ID
285224
Gene Name
DNAJB8 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 3: 128463594 - 128472317 on Build GRCh38
UniGene ID
Hs.683815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037890.1 3 1780
GenBank mRNA AK097457.1 - 3 1781


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000471626.1 - 3 1780 Chr. 3 - 128463594 - 128472317 [+]

View Full Product Details