Gene Transcripts
- Gene Symbol
- DNAJB8-AS1
- Entrez Gene ID
- 285224
- Gene Name
- DNAJB8 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 128463594 - 128472317 on Build GRCh38
- UniGene ID
- Hs.683815
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037890.1 |
|
3 |
1277 |
|
GenBank mRNA |
AK097457.1 |
- |
3 |
1278 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000471626.1 |
- |
3 |
1277 |
|
Chr. 3 - 128463594 - 128472317 [+] |
View Full Product Details